siRNA Details
| Gene Symbol: LINC01541 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038325.1 | 6 | 1314 | |||
| NR_038326.1 | 4 | 929 | ||||
| GenBank mRNA | BX648957.1 | - | 6 | 1315 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01541| Gene Symbol: LINC01541 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038325.1 | 6 | 1314 | |||
| NR_038326.1 | 4 | 929 | ||||
| GenBank mRNA | BX648957.1 | - | 6 | 1315 | ||