siRNA Details
| Gene Symbol: PRR31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005266138.4 | XP_005266195.1 | 1 | 454 | ||
| XM_011519282.2 | XP_011517584.1 | 1 | 454 | |||
| GenBank mRNA | BI825966.1 | - | 1 | 104 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PRR31| Gene Symbol: PRR31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005266138.4 | XP_005266195.1 | 1 | 454 | ||
| XM_011519282.2 | XP_011517584.1 | 1 | 454 | |||
| GenBank mRNA | BI825966.1 | - | 1 | 104 | ||