siRNA Details
| Gene Symbol: LOC101929305 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK022893.1 | - | 2 | 1206 | ||
| BC016656.1 | - | 2 | 1148 | |||
| BC037533.2 | - | 2 | 1203 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101929305| Gene Symbol: LOC101929305 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK022893.1 | - | 2 | 1206 | ||
| BC016656.1 | - | 2 | 1148 | |||
| BC037533.2 | - | 2 | 1203 | |||