siRNA Details
| Gene Symbol: LOC105370622 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135190.1 | 2 | 806 | |||
| GenBank mRNA | BC039357.1 | - | 2 | 806 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105370622| Gene Symbol: LOC105370622 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135190.1 | 2 | 806 | |||
| GenBank mRNA | BC039357.1 | - | 2 | 806 | ||