siRNA Details
Gene Symbol: LOC105370622 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135190.1 | 2 | 1828 | |||
GenBank mRNA | BC039357.1 | - | 2 | 1828 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC105370622 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135190.1 | 2 | 1828 | |||
GenBank mRNA | BC039357.1 | - | 2 | 1828 |