siRNA Details
| Gene Symbol: LOC105376480 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134497.1 | 2 | 242 | |||
| GenBank mRNA | BX114033.1 | - | 2 | 242 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105376480| Gene Symbol: LOC105376480 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134497.1 | 2 | 242 | |||
| GenBank mRNA | BX114033.1 | - | 2 | 242 | ||