siRNA Details
| Gene Symbol: LOC284930 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_122046.1 | 6 | 787 | |||
| GenBank mRNA | AK093107.1 | - | 6 | 758 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC284930| Gene Symbol: LOC284930 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_122046.1 | 6 | 787 | |||
| GenBank mRNA | AK093107.1 | - | 6 | 758 | ||