siRNA Details
| Gene Symbol: LOC101927356 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110076.1 | 5 | 580 | |||
| NR_110077.1 | 4 | 496 | ||||
| GenBank mRNA | AK055932.1 | - | 5 | 580 | ||
| BM468380.1 | - | 4 | 493 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101927356| Gene Symbol: LOC101927356 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110076.1 | 5 | 580 | |||
| NR_110077.1 | 4 | 496 | ||||
| GenBank mRNA | AK055932.1 | - | 5 | 580 | ||
| BM468380.1 | - | 4 | 493 | |||