siRNA Details
| Gene Symbol: LINC00673 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_036488.1 | 1 | 569 | |||
| NR_137280.1 | 1 | 571 | ||||
| NR_137281.1 | 1 | 571 | ||||
| GenBank mRNA | AK000477.1 | - | 1 | 569 | ||
| AK129763.1 | - | 2 | 304 | |||
| KR778895.1 | - | 1 | 569 | |||
| KR778896.1 | - | 1 | 569 | |||
| KR778900.1 | - | 1 | 569 | |||

LINC00673
Genomic Map