siRNA Details
| Gene Symbol: MRVI1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034093.2 | 3 | 599 | |||
| NR_034094.2 | 3 | 561 | ||||
| NR_046374.1 | 5 | 838 | ||||
| NR_046375.1 | 4 | 738 | ||||
| GenBank mRNA | BC028123.1 | - | 3 | 525 | 5167345 | |
| DB071465.1 | - | 4 | 523 | |||

MRVI1-AS1
Genomic Map