siRNA Details
| Gene Symbol: MRVI1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034093.2 | 2 | 330 | |||
| NR_034094.2 | 2 | 330 | ||||
| NR_046374.1 | 4 | 569 | ||||
| NR_046375.1 | 3 | 469 | ||||
| GenBank mRNA | BC028123.1 | - | 2 | 294 | 5167345 | |
| BI520309.1 | - | 4 | 508 | |||
| BU569149.1 | - | 3 | 396 | |||
| BU570065.1 | - | 2 | 238 | |||
| DB071465.1 | - | 2 | 333 | |||
| DB454301.1 | - | 2 | 283 | |||

MRVI1-AS1
Genomic Map