siRNA Details
| Gene Symbol: LINC01079 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BI715219.1 | - | 1 | 156 | ||
| BM503984.1 | - | 1 | 150 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01079| Gene Symbol: LINC01079 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BI715219.1 | - | 1 | 156 | ||
| BM503984.1 | - | 1 | 150 | |||