siRNA Details
| Gene Symbol: LOC101929608 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109878.1 | 1 | 13 | |||
| GenBank mRNA | AK310670.1 | - | 1 | 13 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DC409076.1 | - | 1 | 13 | Chr. 20 - 21612554 - 21703585 [-] on Build GRCh38 | |

LOC101929608
Genomic Map