siRNA Details
| Gene Symbol: PLA2G4E-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120334.1 | 2, 3 | 384 | |||
| GenBank mRNA | BC043275.2 | - | 2, 3 | 383 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PLA2G4E-AS1| Gene Symbol: PLA2G4E-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120334.1 | 2, 3 | 384 | |||
| GenBank mRNA | BC043275.2 | - | 2, 3 | 383 | ||