siRNA Details
| Gene Symbol: LOC105371925 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135654.1 | 3 | 987 | |||
| GenBank mRNA | AL832593.1 | - | 3 | 987 | ||
| BI906297.1 | - | 3 | 352 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105371925| Gene Symbol: LOC105371925 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135654.1 | 3 | 987 | |||
| GenBank mRNA | AL832593.1 | - | 3 | 987 | ||
| BI906297.1 | - | 3 | 352 | |||