siRNA Details
| Gene Symbol: ARRDC3-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027435.1 | 3 | 1880 | |||
| GenBank mRNA | BC043415.2 | - | 3 | 1869 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ARRDC3-AS1| Gene Symbol: ARRDC3-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027435.1 | 3 | 1880 | |||
| GenBank mRNA | BC043415.2 | - | 3 | 1869 | ||