siRNA Details
| Gene Symbol: OIP5-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK092575.1 | - | 1 | 2723 | ||
| BC016329.1 | - | 1 | 1155 | |||
| BC037888.1 | - | 1 | 3861 | |||
| BC047377.1 | - | 1 | 2877 | |||
| BX648343.1 | - | 1 | 4994 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OIP5-AS1| Gene Symbol: OIP5-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK092575.1 | - | 1 | 2723 | ||
| BC016329.1 | - | 1 | 1155 | |||
| BC037888.1 | - | 1 | 3861 | |||
| BC047377.1 | - | 1 | 2877 | |||
| BX648343.1 | - | 1 | 4994 | |||