siRNA Details
| Gene Symbol: LINC01123 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046110.1 | 2 | 1275 | |||
| NR_046111.1 | 2 | 1275 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01123| Gene Symbol: LINC01123 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046110.1 | 2 | 1275 | |||
| NR_046111.1 | 2 | 1275 | ||||