siRNA Details
| Gene Symbol: LINC01123 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046110.1 | 2 | 699 | |||
| NR_046111.1 | 2 | 699 | ||||
| GenBank mRNA | AK056084.1 | - | 2 | 616 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01123| Gene Symbol: LINC01123 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046110.1 | 2 | 699 | |||
| NR_046111.1 | 2 | 699 | ||||
| GenBank mRNA | AK056084.1 | - | 2 | 616 | ||