siRNA Details
| Gene Symbol: LINC01600 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_131168.1 | 3 | 1220 | |||
| GenBank mRNA | AK056496.1 | - | 3 | 1220 | ||
| BC104007.1 | - | 1 | 1004 | 40024375 | ||
| BC104008.1 | - | 1 | 1004 | 40024376 | ||
| BC104009.1 | - | 1 | 1004 | 40024379 | ||

LINC01600
Genomic Map