siRNA Details
| Gene Symbol: LINC01060 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033869.1 | 3, 4 | 253 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BX090209.1 | - | 3, 4 | 240 | Chr. 4 - 188455591 - 188601809 [+] on Build GRCh38 | |

LINC01060
Genomic Map