siRNA Details
| Gene Symbol: MIF-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038911.1 | 3 | 678 | |||
| GenBank mRNA | BC036909.1 | - | 3 | 678 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BI830107.1 | - | 3 | 679 | Chr. 22 - 23895180 - 23898931 [-] on Build GRCh38 | |

MIF-AS1
Genomic Map