siRNA Details
Gene Symbol: PTPRD-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_121599.1 | 3 | 618 | |||
NR_121600.1 | 2 | 536 | ||||
GenBank mRNA | AK094342.1 | - | 2 | 1246 | ||
DB495850.1 | - | 2 | 412 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DB472955.1 | - | 2 | 405 | Chr. 9 - 8858149 - 8861062 [+] on Build GRCh38 |
