siRNA Details
| Gene Symbol: LOC105370333 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_132422.1 | 1 | 294 | |||
| GenBank mRNA | AV692313.1 | - | 1 | 294 | ||
| BG033002.1 | - | 1 | 280 | |||
| CD248433.1 | - | 1 | 231 | |||
| CF454479.1 | - | 1 | 241 | |||
| CX784348.1 | - | 1 | 199 | |||

LOC105370333
Genomic Map