siRNA Details
| Gene Symbol: LOC105370333 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_132422.1 | 2 | 763 | |||
| GenBank mRNA | CX784348.1 | - | 2 | 668 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105370333| Gene Symbol: LOC105370333 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_132422.1 | 2 | 763 | |||
| GenBank mRNA | CX784348.1 | - | 2 | 668 | ||