siRNA Details
| Gene Symbol: LOC101928132 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135052.1 | 2 | 624 | |||
| GenBank mRNA | BC015849.1 | - | 2 | 628 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101928132| Gene Symbol: LOC101928132 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135052.1 | 2 | 624 | |||
| GenBank mRNA | BC015849.1 | - | 2 | 628 | ||