siRNA Details
| Gene Symbol: KLF17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC031250.1 | - | 7 | 2004 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KLF17| Gene Symbol: KLF17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC031250.1 | - | 7 | 2004 | ||