siRNA Details
| Gene Symbol: SIRPG-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110090.1 | 3 | 2654 | |||
| GenBank mRNA | AK093519.1 | - | 3 | 2661 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SIRPG-AS1| Gene Symbol: SIRPG-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110090.1 | 3 | 2654 | |||
| GenBank mRNA | AK093519.1 | - | 3 | 2661 | ||