siRNA Details
| Gene Symbol: LOC284294 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033881.1 | 10 | 1389 | |||
| GenBank mRNA | BC036306.1 | - | 10 | 1388 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC284294| Gene Symbol: LOC284294 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033881.1 | 10 | 1389 | |||
| GenBank mRNA | BC036306.1 | - | 10 | 1388 | ||