siRNA Details
| Gene Symbol: LINC01350 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110793.1 | 3 | 271 | |||
| GenBank mRNA | AF508906.1 | - | 3 | 271 | ||
| AF510427.1 | - | 3 | 261 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CR741638.1 | - | 3 | 263 | Chr. 1 - 185558372 - 185628480 [-] on Build GRCh38 | |

LINC01350
Genomic Map