siRNA Details
Gene Symbol: LINC01350 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110793.1 | 3, 4 | 311 | |||
GenBank mRNA | AF508906.1 | - | 3, 4 | 311 | ||
AF510427.1 | - | 3, 4 | 301 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CR741638.1 | - | 3, 4 | 303 | Chr. 1 - 185558372 - 185628480 [-] on Build GRCh38 |
