siRNA Details
| Gene Symbol: LINC00630 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038988.1 | 7 | 785 | |||
| GenBank mRNA | BC032741.1 | - | 3 | 287 | ||
| BC040959.1 | - | 7 | 773 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00630| Gene Symbol: LINC00630 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038988.1 | 7 | 785 | |||
| GenBank mRNA | BC032741.1 | - | 3 | 287 | ||
| BC040959.1 | - | 7 | 773 | |||