siRNA Details
| Gene Symbol: LINC01094 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038303.1 | 5 | 1994 | |||
| NR_038304.1 | 4 | 1922 | ||||
| NR_038305.1 | 4 | 1900 | ||||
| NR_038306.1 | 3 | 1828 | ||||
| NR_038307.1 | 3 | 1813 | ||||
| NR_038308.1 | 3 | 1774 | ||||
| GenBank mRNA | AK057213.1 | - | 3 | 1606 | ||
| AK124062.1 | - | 4 | 1728 | |||
| AK130099.1 | - | 5 | 1803 | |||
| BC040182.2 | - | 3 | 1551 | |||
| BC044613.1 | - | 3 | 1829 | |||

LINC01094
Genomic Map