siRNA Details
Gene Symbol: LINC01094 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038303.1 | 5 | 1112 | |||
NR_038304.1 | 4 | 1040 | ||||
NR_038305.1 | 4 | 1018 | ||||
NR_038306.1 | 3 | 946 | ||||
NR_038307.1 | 3 | 931 | ||||
NR_038308.1 | 3 | 892 | ||||
GenBank mRNA | AK057213.1 | - | 3 | 724 | ||
AK124062.1 | - | 4 | 846 | |||
AK130099.1 | - | 5 | 920 | |||
BC040182.2 | - | 3 | 669 | |||
BC044613.1 | - | 3 | 946 |
