siRNA Details
| Gene Symbol: LOC101929210 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125922.1 | 6 | 381 | |||
| GenBank mRNA | BC014345.1 | - | 6 | 384 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101929210| Gene Symbol: LOC101929210 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125922.1 | 6 | 381 | |||
| GenBank mRNA | BC014345.1 | - | 6 | 384 | ||