siRNA Details
| Gene Symbol: MACROD2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_072988.2 | 2 | 1216 | |||
| GenBank mRNA | BC039528.1 | - | 2 | 1213 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MACROD2-AS1| Gene Symbol: MACROD2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_072988.2 | 2 | 1216 | |||
| GenBank mRNA | BC039528.1 | - | 2 | 1213 | ||