siRNA Details
| Gene Symbol: MACROD2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_072988.2 | 2 | 952 | |||
| GenBank mRNA | BC039528.1 | - | 2 | 949 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BM564091.1 | - | 2 | 968 | Chr. 20 - 14905116 - 14929524 [-] on Build GRCh38 | |

MACROD2-AS1
Genomic Map