siRNA Details
Gene Symbol: LINC01494 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110238.1 | 3 | 404 | |||
GenBank mRNA | BC038542.1 | - | 3 | 403 | ||
BI488698.1 | - | 3 | 402 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC01494 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110238.1 | 3 | 404 | |||
GenBank mRNA | BC038542.1 | - | 3 | 403 | ||
BI488698.1 | - | 3 | 402 |