siRNA Details
| Gene Symbol: ZNRF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011530438.2 | XP_011528740.1 | 1 | 309 | ||
| XM_017028990.1 | XP_016884479.1 | 1 | 309 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNRF3| Gene Symbol: ZNRF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011530438.2 | XP_011528740.1 | 1 | 309 | ||
| XM_017028990.1 | XP_016884479.1 | 1 | 309 | |||