siRNA Details
| Gene Symbol: LEMD1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038425.1 | 4 | 972 | |||
| GenBank mRNA | AK090497.1 | - | 4 | 972 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LEMD1-AS1| Gene Symbol: LEMD1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038425.1 | 4 | 972 | |||
| GenBank mRNA | AK090497.1 | - | 4 | 972 | ||