siRNA Details
| Gene Symbol: C20orf78 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001242671.2 | NP_001229600.1 | 1 | 107 | ||
| NM_001302818.1 | NP_001289747.1 | 1 | 107 | |||
| GenBank mRNA | BX112808.1 | - | 1 | 57 | ||
| BX334499.2 | - | 1 | 68 | |||
| CR738909.1 | - | 1 | 99 | |||
| HY016900.1 | - | 1 | 65 | |||
| HY050710.1 | - | 1 | 108 | |||

C20orf78
Genomic Map