siRNA Details
Gene Symbol: KCNC4-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046546.1 | 2 | 503 | |||
GenBank mRNA | BX089534.1 | - | 2 | 503 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: KCNC4-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046546.1 | 2 | 503 | |||
GenBank mRNA | BX089534.1 | - | 2 | 503 |