siRNA Details
| Gene Symbol: LOC285389 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_136181.1 | 4 | 2275 | |||
| GenBank mRNA | BC038368.1 | - | 4 | 2275 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC285389| Gene Symbol: LOC285389 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_136181.1 | 4 | 2275 | |||
| GenBank mRNA | BC038368.1 | - | 4 | 2275 | ||