siRNA Details
| Gene Symbol: LOC100288181 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038925.1 | 3 | 2789 | |||
| GenBank mRNA | CR627188.1 | - | 3 | 2790 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100288181| Gene Symbol: LOC100288181 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038925.1 | 3 | 2789 | |||
| GenBank mRNA | CR627188.1 | - | 3 | 2790 | ||