siRNA Details
| Gene Symbol: LOC101927378 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110079.1 | 3 | 2998 | |||
| GenBank mRNA | BX647900.1 | - | 3 | 2999 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101927378| Gene Symbol: LOC101927378 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110079.1 | 3 | 2998 | |||
| GenBank mRNA | BX647900.1 | - | 3 | 2999 | ||