siRNA Details
| Gene Symbol: LOC100996351 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110670.1 | 4 | 1670 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100996351| Gene Symbol: LOC100996351 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110670.1 | 4 | 1670 | |||