siRNA Details
| Gene Symbol: LOC100996351 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110670.1 | 1, 2 | 165 | |||
| GenBank mRNA | AK123032.1 | - | 1, 2 | 165 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DA584105.1 | - | 1, 2 | 165 | Chr. 19 - 3149363 - 3155175 [-] on Build GRCh38 | |

LOC100996351
Genomic Map