siRNA Details
Gene Symbol: TTTY5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_001541.1 | 1 | 84 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AF332236.3 | - | 1 | 84 | Chr. Y - 22296794 - 22298876 [-] on Build GRCh38 |
