siRNA Details
| Gene Symbol: TTTY5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_001541.1 | 2 | 998 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AF332236.3 | - | 2 | 998 | Chr. Y - 22296794 - 22298876 [-] on Build GRCh38 | |

TTTY5
Genomic Map