siRNA Details
Gene Symbol: LOC101929626 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125987.1 | 3 | 299 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BG182305.1 | - | 2 | 287 | Chr. 1 - 46134524 - 46139081 [+] on Build GRCh38 |
